Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

498327

Sigma-Aldrich

Ethyl 2-fluoroacetoacetate

Synonym(s):

2-Fluoro-3-oxobutanoic acid ethyl ester, Ethyl 2-fluoro-3-oxobutanoate, Ethyl 2-fluoro-3-oxobutyrate, NSC 24563

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH(F)CO2C2H5
CAS Number:
Molecular Weight:
148.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

refractive index

n20/D 1.414 (lit.)

bp

183 °C (lit.)

density

1.181 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C(F)C(C)=O

InChI

1S/C6H9FO3/c1-3-10-6(9)5(7)4(2)8/h5H,3H2,1-2H3

InChI key

SHTFQLHOTAJQRJ-UHFFFAOYSA-N

General description

Ethyl 2-fluoroacetoacetate is an α-fluorinated β-keto ester that can be prepared by the fluorination of ethyl acetoacetate.

Application

Ethyl 2-fluoroacetoacetate may be used in the preparation of:
  • δ-keto-β-hydroxy-α-fluoro esters
  • fluorinated [1,2,4]triazolo[1,5-a]pyrimidin-7(4H)-ones
  • fluorinated [1,2,4]triazolo[5,1-c][1,2,4]triazin-4(1H)-ones
  • 4-fluorofarnesol
Reactant for:
  • Michael addition-induced cyclization reaction
  • Asymmetric Mannich reaction
  • Enantioselective organocatalytic conjugate addition

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Green chemistry: synthesis in micro reactors.
Haswell SJ and Watts P.
Green Chemistry, 5(2), 240-249 (2003)
Organocatalytic direct aldol reaction between acetone and α-substituted β-keto esters.
London G, et al.
J. Mol. Catal. A: Chem., 267(1), 98-101 (2007)
Fluorinated [1,2,4] triazolo [1,5-a] pyrimidines and [1,2,4] triazolo [5,1-c][1,2,4] triazines*.
Ulomskiy EN, et al.
Chemistry of Heterocyclic Compounds, 47(9), 1164-1169 (2011)
Fluoroisoprenyl synthesis using ethyl 2-fluoroacetoacetate.
Ortiz de Montellano PR and Vinson WA.
The Journal of Organic Chemistry, 42(11), 2013-2014 (1977)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service