Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

496731

Sigma-Aldrich

Bisphenol F ethoxylate (2 EO/phenol) diacrylate

average Mn ~484

Synonym(s):

Ethoxylated bisphenol F diacrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[H2C=CHCO2(CH2CH2O)nC6H4-4-]2CH2
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

mol wt

average Mn ~484

Quality Level

contains

100 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.541 (lit.)

density

1.15 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

OCCO.OC(=O)C=C.Oc1ccc(Cc2ccc(O)cc2)cc1

InChI

1S/C13H12O2.C3H4O2.C2H6O2/c14-12-5-1-10(2-6-12)9-11-3-7-13(15)8-4-11;1-2-3(4)5;3-1-2-4/h1-8,14-15H,9H2;2H,1H2,(H,4,5);3-4H,1-2H2

InChI key

OTXNLBPVRKDFKX-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service