Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

496480

Sigma-Aldrich

Bis[4-(vinyloxy)butyl] succinate

98%

Synonym(s):

VEctomer® 4030 vinyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CO2(CH2)4OCH=CH2]2
CAS Number:
Molecular Weight:
314.37
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

98%

refractive index

n20/D 1.461 (lit.)

density

1.06 g/mL at 25 °C (lit.)

SMILES string

C=COCCCCOC(=O)CCC(=O)OCCCCOC=C

InChI

1S/C16H26O6/c1-3-19-11-5-7-13-21-15(17)9-10-16(18)22-14-8-6-12-20-4-2/h3-4H,1-2,5-14H2

InChI key

XUEAJYHEEJKSLM-UHFFFAOYSA-N

Legal Information

Product of Morflex, Inc.
VEctomer is a registered trademark of Vertellus Specialties, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service