Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

494577

Sigma-Aldrich

(R)-(−)-2-Amino-1-phenylethanol

97%

Synonym(s):

(R)-α-(Aminomethyl)benzyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCH2CH(C6H5)OH
CAS Number:
Molecular Weight:
137.18
Beilstein:
3196196
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]22/D −39°, c = 2 in ethanol

mp

59-65 °C (lit.)

SMILES string

NC[C@H](O)c1ccccc1

InChI

1S/C8H11NO/c9-6-8(10)7-4-2-1-3-5-7/h1-5,8,10H,6,9H2/t8-/m0/s1

InChI key

ULSIYEODSMZIPX-QMMMGPOBSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service