Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

493880

Sigma-Aldrich

(3aS,7R,7aS)-7,7a-Dihydro-7-hydroxy-2,2-dimethyl-1,3-benzodioxol-4(3aH)-one

0.2 g/mL in 0.1 M phosphate buffer, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12O4
CAS Number:
Molecular Weight:
184.19
MDL number:
UNSPSC Code:
12352103

Assay

98%

concentration

0.2 g/mL in 0.1 M phosphate buffer

pH

8.5(adjusted to)

SMILES string

CC1(C)O[C@H]2[C@H](O)C=CC(=O)[C@H]2O1

Physical form

A suspension in phosphate buffer.

Reconstitution

To recover the pure product from the suspension:
(1)Thaw the frozen suspension and filter the solid. (2) Rinse bottle with a few milliliters of base-washed (aqueous Na2CO3) ethyl acetate. (3) Use the rinsing to wash the solid. (4) Collect solid. (5) To further collect more product from the filtrate: extract filtratewith equal volumes of base-washed (aqueous Na2CO3) ethyl acetate (repeat three times). (6) Dry the ethyl acetate extract over MgSO4. (7) Evaporate the solvent - DO NOT HEAT! (8) Combine solids collected from the suspension and the filtrate. Pure crystals should be stored at -78° C. A suspension of the product in phosphate buffer is stable at 0° C.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service