Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

493317

Sigma-Aldrich

Propane-2,2-d2

98 atom % D

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
$1,660.00

$1,660.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 L
$1,660.00

About This Item

Linear Formula:
CH3CD2CH3
CAS Number:
Molecular Weight:
46.11
MDL number:
UNSPSC Code:
12142200
PubChem Substance ID:
NACRES:
NA.12

$1,660.00


Please contact Customer Service for Availability

Request more information

vapor density

1.5 (vs air)

Quality Level

vapor pressure

8.42 atm ( 21.1 °C)

isotopic purity

98 atom % D

form

gas

bp

−42.1 °C (lit.)

mp

−188 °C (lit.)

mass shift

M+2

SMILES string

[2H]C([2H])(C)C

InChI

1S/C3H8/c1-3-2/h3H2,1-2H3/i3D2

InChI key

ATUOYWHBWRKTHZ-SMZGMGDZSA-N

Related Categories

Packaging

Standard 1 L quantity is packaged in a 450 mL carbon steel lecture bottle with brass CGA 110/180 valve. Nominal gas pressure at 21ºC is 20 psig for 1 L. This pressure is slightly above atmospheric pressure. Care must be taken when extracting this product from the cylinder.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Recommended products

Stainless steel control valves Z146951 or Z187224
PTFE sealing washers GCA inlet size 180 Z146994
PTFE sealing washers CGA inlet size 110 Z146986

Pictograms

FlameGas cylinder

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Gas 1 - Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

-155.2 °F - closed cup

Flash Point(C)

-104.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service