Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

492884

Sigma-Aldrich

Boc-Ala-OH-12C3

99.9 atom % 12C

Synonym(s):

N-(tert-Butoxycarbonyl)-L-alanine-12C3, L-Alanine-12C3, N-t-Boc derivative

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
12CH312CH(NH-Boc)12CO2H
Molecular Weight:
189.18
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

description

13C-depleted

Quality Level

isotopic purity

99.9 atom % 12C

optical activity

[α]20/D -23°, c = 2 in acetic acid

technique(s)

bio NMR: suitable

mp

79-83 °C (lit.)

application(s)

peptide synthesis

functional group

Boc

mass shift

depleted

SMILES string

C[C@H](NC(=O)OC(C)(C)C)C(O)=O

InChI

1S/C8H15NO4/c1-5(6(10)11)9-7(12)13-8(2,3)4/h5H,1-4H3,(H,9,12)(H,10,11)/t5-/m0/s1

InChI key

QVHJQCGUWFKTSE-YFKPBYRVSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huang-Han Chen et al.
Analytica chimica acta, 865, 53-59 (2015-03-04)
Polydimethylsiloxane (PDMS) is widely used for microfabrication and bioanalysis; however, its surface functionalization is limited due to the lack of active functional groups and incompatibility with many solvents. We presented a novel approach for in situ fabrication of cleavable peptide
Keyvan Dastmalchi et al.
Journal of agricultural and food chemistry, 63(30), 6810-6822 (2015-07-15)
The cultivation, storage, and distribution of potato tubers are compromised by mechanical damage and suboptimal healing. To investigate wound-healing progress in cultivars with contrasting russeting patterns, metabolite profiles reported previously for polar tissue extracts were complemented by GC/MS measurements for

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service