Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

490881

Sigma-Aldrich

[3aR-(3aα,5aβ,6aβ,6bα)]-3a,5a,6a,6b-Tetrahydro-2,2-dimethyloxireno[e]-1,3-benzodioxole

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12O3
CAS Number:
Molecular Weight:
168.19
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

optical activity

[α]18/D −56°, c = 1.3 in chloroform

refractive index

n20/D 1.476 (lit.)

bp

79 °C/1.5 mmHg (lit.)

density

1.101 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

[H][C@]12O[C@@]1([H])[C@]3([H])OC(C)(C)O[C@]3([H])C=C2

InChI

1S/C9H12O3/c1-9(2)11-6-4-3-5-7(10-5)8(6)12-9/h3-8H,1-2H3/t5-,6-,7-,8-/m1/s1

InChI key

GAUDXMCZQNKKEF-WCTZXXKLSA-N

Storage Class Code

10 - Combustible liquids not in Storage Class 3

WGK

WGK 3

Flash Point(F)

198.0 °F - closed cup

Flash Point(C)

92.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu-Wen Alvin Huang et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 39(37), 7408-7427 (2019-07-25)
In blood, apolipoprotein E (ApoE) is a component of circulating lipoproteins and mediates the clearance of these lipoproteins from blood by binding to ApoE receptors. Humans express three genetic ApoE variants, ApoE2, ApoE3, and ApoE4, which exhibit distinct ApoE receptor-binding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service