Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

487368

Sigma-Aldrich

4-(Benzyloxy)phenyl isocyanate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OC6H4NCO
CAS Number:
Molecular Weight:
225.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

bp

129 °C/0.2 mmHg (lit.)

mp

60-62 °C (lit.)

storage temp.

2-8°C

SMILES string

O=C=Nc1ccc(OCc2ccccc2)cc1

InChI

1S/C14H11NO2/c16-11-15-13-6-8-14(9-7-13)17-10-12-4-2-1-3-5-12/h1-9H,10H2

InChI key

HFIHSWMJFCMLJE-UHFFFAOYSA-N

General description

4-(Benzyloxy)phenyl isocyanate, also known as 1-(benzyloxy)-4-isocyanatobenzene, is an organic building block containing an isocyanate group.

Application

4-(Benzyloxy)phenyl isocyanate may be used in the synthesis of 1,3,5-tris(4-benzyloxyphenyl)-1,3,5-triazinane-2,4,6-trione via cyclotrimerization.[1]
It may be used in the synthesis of the following phenyl urea derivatives:[2]
  • 1-[4-(benzyloxy)phenyl]-3-(1-methyl-3-phenyl-1H-pyrazol-5-yl)urea
  • 1-[4-(benzyloxy)phenyl]-3-[3-(4-chlorophenyl)-1-phenyl-1H-pyrazol-5-yl]urea
  • 1-[4-(benzyloxy)phenyl]-3-[3-(2-chlorophenyl)-1-phenyl-1H-pyrazol-5-yl]urea
  • N-[4-[3-[4-(benzyloxy)phenyl]ureido]phenyl]nicotinamide
  • 1-[4-(benzyloxy)phenyl]-3-(1-nicotinoylindolin-5-yl)urea
  • 4-[3-[4-(benzyloxy)phenyl]ureido]-N-phenylbenzamide

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gustav Boije af Gennäs et al.
ChemMedChem, 6(9), 1680-1692 (2011-07-02)
In anaplastic large-cell lymphomas, chromosomal translocations involving the kinase domain of anaplastic lymphoma kinase (ALK), generally fused to the 5' part of the nucleophosmin gene, produce highly oncogenic ALK fusion proteins that deregulate cell cycle, apoptosis, and differentiation in these
Bhasker Bantu et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(13), 3103-3109 (2009-02-13)
Catalytic rivals: Both CO(2)-protected tetrahydropyrimidin-2-ylidene-based N-heterocyclic carbenes (NHCs) and Sn(II)-1,3-dimesitylimidazol-2-ylidene, as well as Sn(II)-1,3-dimesitylimidazolin-2-ylidene complexes (example displayed), have been identified as truly latent catalysts for polyurethane (PUR) synthesis rivaling all existing systems both in activity and latency.A series of CO(2)-protected

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service