Skip to Content
MilliporeSigma
All Photos(1)

Documents

486620

Sigma-Aldrich

Dicarbonyl(pentamethylcyclopentadienyl)chromium(V) dimer

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H30Cr2O4
CAS Number:
Molecular Weight:
486.48
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

97%

reaction suitability

core: chromium
reagent type: catalyst

mp

~220 °C (dec.) (lit.)

SMILES string

[Cr].[Cr].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].C[C]1[C](C)[C](C)[C](C)[C]1C.C[C]2[C](C)[C](C)[C](C)[C]2C

InChI

1S/2C10H15.4CO.2Cr/c2*1-6-7(2)9(4)10(5)8(6)3;4*1-2;;/h2*1-5H3;;;;;;

InChI key

DYOFVXQOZUEOEJ-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service