Skip to Content
MilliporeSigma
All Photos(2)

Documents

484415

Sigma-Aldrich

Propylene glycol butyl ether

≥99%

Synonym(s):

DOWANOL® PnB

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3OC3H6OH
CAS Number:
Molecular Weight:
132.20
EC Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

Assay

≥99%

composition

(mixture of isomers)

refractive index

n20/D 1.416 (lit.)

bp

165-175 °C (lit.)

density

0.885 g/mL at 25 °C

InChI

1S/C7H16O2/c1-3-4-5-9-6-7(2)8/h7-8H,3-6H2,1-2H3

InChI key

RWNUSVWFHDHRCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Product of Dow Chemical
DOWANOL is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mitchell Tiessen et al.
Journal of chromatographic science, 59(2), 103-111 (2020-11-05)
Paints are a common form of physical evidence encountered at crime scenes. This research presents an optimized method for the untargeted analysis of volatile organic compounds (VOCs) in spray paint using solid-phase microextraction-gas chromatography-mass spectrometry (SPME-GC-MS). The presence and persistence
W B Gibson et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 13(3), 359-365 (1989-10-01)
The developmental toxicity (embryofetotoxicity or teratogenicity) and maternal toxicity of butoxypropanol, a propylene glycol ether, was evaluated in New Zealand White rabbits. Dosages of 0 (water), 10, 40 or 100 mg/kg/day, selected on the basis of the water solubility of
Formaldehyde, 2-butoxyethanol and 1-tert-butoxypropan-2-ol.
IARC monographs on the evaluation of carcinogenic risks to humans, 88, 1-478 (2007-03-21)
Vincent James Cogliano et al.
Environmental health perspectives, 113(9), 1205-1208 (2005-09-06)
An international, interdisciplinary working group of expert scientists met in June 2004 to develop IARC Monographs on the Evaluation of the Carcinogenic Risk of Chemicals to Humans (IARC Monographs) on formaldehyde, 2-butoxyethanol, and 1-tert-butoxy-2-propanol. Each IARC Monograph includes a critical
E R Kennedy et al.
Journal of chromatography, 522, 303-313 (1990-11-28)
The applicability of multidimensional gas chromatography-mass spectrometry to the analysis of five glycol ethers in air was demonstrated. Air samples were collected on charcoal tubes and desorbed with 5% methanol in methylene chloride as is described in method 1403 of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service