Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

484199

Sigma-Aldrich

Cyclopentadienyl(fluorene)iron(II) hexafluorophosphate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H15F6FeP
CAS Number:
Molecular Weight:
432.12
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

98%

reaction suitability

reagent type: catalyst
core: iron

mp

163 °C (dec.) (lit.)

SMILES string

[Fe+].[CH]1[CH][CH][CH][CH]1.F[P-](F)(F)(F)(F)F.[CH]2[C]3C=CC=C[C]3[C]4C=CC=C[C]24

InChI

1S/C13H9.C5H5.F6P.Fe/c1-3-7-12-10(5-1)9-11-6-2-4-8-13(11)12;1-2-4-5-3-1;1-7(2,3,4,5)6;/h1-9H;1-5H;;/q;;-1;+1

InChI key

OMTIHTHOHAUYEQ-UHFFFAOYSA-N

Application

  • Catalytic photoinitiator for radical and cationic photochemical polymerization

Reactant for:
  • Solid-state synthesis of carbon nanostructures
  • Synthesis of fulvalene-bridged bimetallic compounds based on Pauson-Khand chemistry

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service