Skip to Content
MilliporeSigma
All Photos(1)

Documents

483796

Sigma-Aldrich

D-(−)-Tartaric acid

99%

Synonym(s):

(2S,3S)-(−)-Tartaric acid, D-Threaric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH(OH)CH(OH)CO2H
CAS Number:
Molecular Weight:
150.09
Beilstein:
1725145
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

description

synthetic

Assay

99%

optical activity

[α]20/D −12°, c = 20 in H2O

optical purity

ee: 97% (GLC)

mp

172-174 °C (lit.)

SMILES string

O[C@@H]([C@H](O)C(O)=O)C(O)=O

InChI

1S/C4H6O6/c5-1(3(7)8)2(6)4(9)10/h1-2,5-6H,(H,7,8)(H,9,10)/t1-,2-/m0/s1

InChI key

FEWJPZIEWOKRBE-LWMBPPNESA-N

Looking for similar products? Visit Product Comparison Guide

Application

D-(-)-Tartaric acid may be used in the preparation of enantiospecific homochiral cis-4-formyl β-lactams. It may also be used as a starting material in the synthesis of D-erythro-sphingosine and L-lyxo-phytosphingosine.

Other Notes

Unnatural isomer

replaced by

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An alternative synthesis of d-erythro-sphingosine and l-lyxo-phytosphingosine.
Nakamura T and Shiozaki M.
Tetrahedron Letters, 40(51), 9063-9064 (1999)
Efficient Asymmetric Synthesis of cis-4-Formyl-?-Lactams from L-(+)-Tartaric Acid.
Jayaraman M, et al.
The Journal of Organic Chemistry, 59(4), 932-934 (1994)
Hyo-Hee Kim et al.
Food science and biotechnology, 26(5), 1343-1347 (2018-09-29)
Isomalto-oligosaccharide (IMO) syrup was prepared from organic rice, in which Lactobacillus plantarum as a starter was inoculated to raise its purity and also produce a fermented rice beverage. Of the five strains of lactic acid bacteria tested, L. plantarum was
J B Olivato et al.
Carbohydrate polymers, 92(2), 1705-1710 (2013-02-13)
Tartaric acid (TA), a dicarboxylic acid, can act as a compatibiliser in starch/polyester blends. A mixture design was proposed to evaluate the effect of TA on the properties of starch/poly (butylene adipate co-terephthalate) (PBAT) blown films plasticised with glycerol. The
Mrinal Kanti Bain et al.
Carbohydrate polymers, 91(2), 529-536 (2012-11-06)
Gelation temperature of MC was reduced from 59°C to 54°C with the addition of 10% PEG. Sodium tartrate (NaT) and sodium citrate (NaC) were added to the MC-PEG solution to further reduce the gelation temperature close to physiological temperature. Different

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service