Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

483168

Sigma-Aldrich

Hexaphenylcyclotrisiloxane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H30O3Si3
CAS Number:
Molecular Weight:
594.88
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

bp

300 °C/1 mmHg (lit.)

mp

184-188 °C (lit.)

SMILES string

O1[Si](O[Si](O[Si]1(c2ccccc2)c3ccccc3)(c4ccccc4)c5ccccc5)(c6ccccc6)c7ccccc7

InChI

1S/C36H30O3Si3/c1-7-19-31(20-8-1)40(32-21-9-2-10-22-32)37-41(33-23-11-3-12-24-33,34-25-13-4-14-26-34)39-42(38-40,35-27-15-5-16-28-35)36-29-17-6-18-30-36/h1-30H

InChI key

VCYDUTCMKSROID-UHFFFAOYSA-N

Other Notes

Contains other phenylcyclosiloxanes

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyungkuk Koh et al.
Polymers, 13(4) (2021-02-13)
In this study, a fast curable polysiloxane-silphenylene hybrimer (PSH) was synthesized by the nonhydrolytic sol-gel condensation of phenyl-vinyl-oligosiloxane (PVO) and tris(dimethylhydrosilyl)benzene (TDMSB) under a Pt catalyst to investigate its optical property and thermal stability. The combination of PVO and tripod-type

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service