Skip to Content
MilliporeSigma
All Photos(1)

Documents

482684

Sigma-Aldrich

ArgoGel−MB-CHO

120-230 μm particle size

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

product line

ArgoPore

crosslinking

1 % cross-linked

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis
reactivity: amine reactive

extent of labeling

0.35-0.45 mmol typical aldehyde loading per g

particle size

120-230 μm

functional group

aldehyde

storage temp.

2-8°C

SMILES string

[H]C(=O)c1ccc(O)cc1OC.CC(COCCO)(COCCO)Cc2ccc(C=C)cc2

Legal Information

ArgoGel is a trademark of Argonaut Technologies, Inc.
ArgoPore is a trademark of Argonaut Technologies, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service