Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

482269

Sigma-Aldrich

Sodium cobalticarborane

95%

Synonym(s):

Sodium bis(undecahydro-7,8-dicarbaundecaborato)cobaltate, Sodium bis-1,2-dicarbollylcobaltate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H22B18CoNa
CAS Number:
Molecular Weight:
346.74
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

mp

>300 °C (lit.)

SMILES string

[Na+].[Co-].B1BBBBC2BC2BBBB1.B3BBBBC4BC4BBBB3

InChI

1S/2C2H12B10.Co.Na/c2*3-1-2(3)5-7-9-11-12-10-8-6-4-1;;/h2*1-12H;;/q;;-1;+1

InChI key

WUOJUVSOTXCWMB-UHFFFAOYSA-N

Application

Sodium cobalticarborane was used in a comparitive study against ethylene glycol functionalized polystyrene microspheres (P-EG) based electrodes.1

Packaging

Packaged in glass bottles

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potentiometric Response Characteristics of
Membrane-Based Cs+-Selective Electrodes Containing
Ionophore-Functionalized Polymeric Microspheres, SAGE-Hindawi Access to Research
Peper S and Gonczy C
International Journal of Electrochemistry (2011)
Christopher J Traverse et al.
Scientific reports, 7(1), 16399-16399 (2017-11-29)
Organic molecular salts are an emerging and highly tunable class of materials for organic and transparent photovoltaics. In this work, we demonstrate novel phenyl borate and carborane-based anions paired with a near-infrared (NIR)-selective heptamethine cation. We further explore the effects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service