Skip to Content
MilliporeSigma
All Photos(1)

Documents

482110

Sigma-Aldrich

Methyl cyclopropanecarboxylate

≥99%

Synonym(s):

MCPC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H5CO2CH3
CAS Number:
Molecular Weight:
100.12
Beilstein:
2039388
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥99%

refractive index

n20/D 1.419 (lit.)

bp

119 °C/764 mmHg (lit.)

density

0.985 g/mL at 25 °C (lit.)

SMILES string

COC(=O)C1CC1

InChI

1S/C5H8O2/c1-7-5(6)4-2-3-4/h4H,2-3H2,1H3

InChI key

PKAHQJNJPDVTDP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

64.4 °F - closed cup

Flash Point(C)

18 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Didier Ndeh et al.
Nature communications, 11(1), 646-646 (2020-02-02)
The human gut microbiota (HGM), which is critical to human health, utilises complex glycans as its major carbon source. Glycosaminoglycans represent an important, high priority, nutrient source for the HGM. Pathways for the metabolism of various glycosaminoglycan substrates remain ill-defined.
Lucy Yang et al.
Chest, 151(3), 555-563 (2016-11-26)
Smoking and COPD are risk factors for cardiovascular disease, and the pathogenesis may involve endothelial dysfunction. We tested the hypothesis that endothelium-derived epoxyeicosatrienoic acid (EET)-mediated endothelial function is impaired in patients with COPD and that a novel soluble epoxide hydrolase
Orna Rosenthal et al.
Journal of neuroengineering and rehabilitation, 14(1), 127-127 (2017-12-07)
Chronic upper limb motor impairment is a common outcome of stroke. Therapeutic training can reduce motor impairment. Recently, a growing interest in evaluating motor training provided by robotic assistive devices has emerged. Robot-assisted therapy is attractive because it provides a
Musa Basseer Sami et al.
Journal of psychopharmacology (Oxford, England), 32(8), 825-849 (2018-03-30)
A substantial body of credible evidence has accumulated that suggest that cannabis use is an important potentially preventable risk factor for the development of psychotic illness and its worse prognosis following the onset of psychosis. Here we summarize the relevant
Lara Al-Olabi et al.
The Journal of clinical investigation, 128(4), 1496-1508 (2018-02-21)
Sporadic vascular malformations (VMs) are complex congenital anomalies of blood vessels that lead to stroke, life-threatening bleeds, disfigurement, overgrowth, and/or pain. Therapeutic options are severely limited, and multidisciplinary management remains challenging, particularly for high-flow arteriovenous malformations (AVM). To investigate the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service