Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

481459

Sigma-Aldrich

Rubidium dihydrogenarsenate

99.9% trace metals basis

Synonym(s):

Dihydrogen rubidium arsenate, Rubidium dihydrogen arsenate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
RbH2AsO4
CAS Number:
Molecular Weight:
226.40
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

99.9% trace metals basis

form

solid

reaction suitability

reagent type: catalyst
core: arsenic

mp

268 °C (lit.)

SMILES string

[Rb+].O[As](O)([O-])=O

InChI

1S/AsH3O4.Rb/c2-1(3,4)5;/h(H3,2,3,4,5);/q;+1/p-1

InChI key

DGQWDYBVPGQHSA-UHFFFAOYSA-M

Application

A new iron(III) arsenate, Rb2Fe2O(AsO4)2, with a novel sheet structure, was synthesized from a solution of the corresponding RbH2AsO4.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rei-Shyang Chang et al.
Inorganic chemistry, 36(16), 3410-3413 (1997-07-30)
Two new iron(III) arsenates K(2)Fe(2)O(AsO(4))(2) (1) and Rb(2)Fe(2)O(AsO(4))(2) (2) have been synthesized and characterized by single-crystal X-ray diffraction, magnetic susceptibilities, and Mossbauer spectroscopy. The two compounds are isostructural and exhibit a sheet structure consisting of infinite slabs of edge-sharing FeO(6)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service