Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

481157

Sigma-Aldrich

Tetraamminepalladium(II) bromide

99.99% trace metals basis

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$329.00

$329.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$329.00

About This Item

Linear Formula:
Pd(NH3)4Br2
CAS Number:
Molecular Weight:
334.35
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

$329.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

for analytical purposes

Assay

99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: palladium

mp

>150 °C (dec.) (lit.)

SMILES string

N.N.N.N.Br[Pd]Br

InChI

1S/2BrH.4H3N.Pd/h2*1H;4*1H3;/q;;;;;;+2/p-2

InChI key

BLPSMPASLSGGOC-UHFFFAOYSA-L

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service