Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

480509

Sigma-Aldrich

Methyl 5-nitro-2-furoate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO5
CAS Number:
Molecular Weight:
171.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

Assay

98%

mp

78-82 °C (lit.)

SMILES string

COC(=O)c1ccc(o1)[N+]([O-])=O

InChI

1S/C6H5NO5/c1-11-6(8)4-2-3-5(12-4)7(9)10/h2-3H,1H3

InChI key

UTLKCGPAJUYGOM-UHFFFAOYSA-N

General description

Methyl 5-nitro-2-furoate is a nitrofuran derivative. Its density, freezing point and refractive index have been evaluated.[1] The reduction of methyl 5-nitro-2-furoate using milk xanthine oxidase has been reported to form methyl 5-hydroxylamino-2-furoate and methyl 5-amino-2-furoate.[2] The utility of methyl 5-nitro-2-furoate as a matrix compound for matrix assisted ionization vacuum (MAIV) has been assessed using bovine insulin as an analyte.[3]

Application

Methyl 5-nitro-2-furoate may be used in the synthesis of 5-nitro-2-furoylhydrazine[4] and 5-nitro-2-furamide.[5]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaws CL.
The Yaws Handbook of Physical Properties for Hydrocarbons and Chemicals, 97-97 (2015)
Reduction of methyl 5-nitro-2-furoate by xanthine oxidase: an evidence for hydroxylaminofuran formation.
H Yamada et al.
Chemical & pharmaceutical bulletin, 30(7), 2647-2650 (1982-07-01)
Synthesis and antimicrobial activity of methyl 5-nitro-3,4-diphenylfuran-2-carboxylate and related compounds.
Kuo SC, et al.
Chemical & Pharmaceutical Bulletin, 29(3), 635-645 (1981)
Magic matrices for ionization in mass spectrometry.
Trimpin S, et al.
International Journal of Mass Spectrometry, 377, 532-545 (2015)
H Yamada et al.
Archives of biochemistry and biophysics, 232(1), 234-239 (1984-07-01)
After methyl 5-nitro-2-furoate was incubated with milk xanthine oxidase, three reduction products were isolated from the incubation mixture. Among them, two reduction products were new types of nitrofuran metabolites, i.e., metabolites 1 and 2 were identified as the dihydroxyhydrazine derivative

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service