Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

480215

Sigma-Aldrich

1,8-Diazabicyclo[5.4.0]undec-7-ene, compound with phenol (1:1)

Synonym(s):

1,8-Diazabicyclo[5.4.0]undec-7-ene phenol salt, DBU phenol salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H22N2O
CAS Number:
Molecular Weight:
246.35
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

density

1.062 g/mL at 25 °C (lit.)

SMILES string

Oc1ccccc1.C2CCN3CCCN=C3CC2

InChI

1S/C9H16N2.C6H6O/c1-2-5-9-10-6-4-8-11(9)7-3-1;7-6-4-2-1-3-5-6/h1-8H2;1-5,7H

InChI key

MFYNHXMPPRNECN-UHFFFAOYSA-N

Application

Gelling catalyst for foams, coatings and adhesives.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service