Skip to Content
MilliporeSigma
All Photos(1)

Documents

480010

Sigma-Aldrich

Potassium ferrate(VI)

97%

Synonym(s):

Dipotassium tetraoxoferrate(VI), Potassium ferrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2FeO4
CAS Number:
Molecular Weight:
198.04
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

97%

form

powder

reaction suitability

reagent type: catalyst
core: iron

mp

>400 °C (lit.)

SMILES string

[K+].[K+].[O-][Fe]([O-])(=O)=O

InChI

1S/Fe.2K.4O/q;2*+1;;;2*-1

InChI key

ICBFRPYDFGVTEQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame over circle

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhengwei Zhou et al.
Journal of pharmaceutical and biomedical analysis, 106, 37-45 (2014-07-27)
The performance of ferrate(VI) in treating sulfamethoxazole (SMX), diclofenac (DCF), carbamazepine (CBZ) and bezafibrate (BZF) in test solutions containing the four compounds was investigated. A series of jar-test experiments was performed on a bench-scale at pH 6-9 and at a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service