Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

479810

Sigma-Aldrich

1-Tridecene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10CH=CH2
CAS Number:
Molecular Weight:
182.35
Beilstein:
1744660
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.432 (lit.)

bp

232-233 °C (lit.)

mp

−23 °C (lit.)

density

0.766 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCC=C

InChI

1S/C13H26/c1-3-5-7-9-11-13-12-10-8-6-4-2/h3H,1,4-13H2,2H3

InChI key

VQOXUMQBYILCKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Substance toxicity and danger].
L V Mel'nikova
Gigiena i sanitariia, (1)(1), 94-94 (1981-01-01)
Qing-He Zhang et al.
Journal of chemical ecology, 30(9), 1849-1870 (2004-12-14)
Gas chromatographic-electroantennographic detection (G3C-EAD) experiments showed that antennae of males and females of the goldeneyed lacewing, Chrysopa oculata Say (Co. = Chrysopa), consistently responded to four compounds extracted from the abdominal cuticle of males:nonanal, nonanol, nonanoic acid, and (1R*,2S*,5R*,8R*)-iridodial. These
Sven Geiselhardt et al.
Die Naturwissenschaften, 95(3), 247-251 (2007-09-28)
Males of the genus Parastizopus (Coleoptera: Tenebrionidae) exhibit a special pheromone-emitting behaviour. They do a headstand, expose the aedeagus and remain in this posture for a few seconds. The pheromone emitted by P. transgariepinus was collected by solid-phase micro-extraction (100

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service