Skip to Content
MilliporeSigma
All Photos(1)

Documents

479667

Sigma-Aldrich

3-Isopropylphenol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHC6H4OH
CAS Number:
Molecular Weight:
136.19
Beilstein:
2040880
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.526 (lit.)

bp

228 °C (lit.)

mp

25 °C (lit.)

density

0.994 g/mL at 25 °C (lit.)

SMILES string

CC(C)c1cccc(O)c1

InChI

1S/C9H12O/c1-7(2)8-4-3-5-9(10)6-8/h3-7,10H,1-2H3

InChI key

VLJSLTNSFSOYQR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Isopropylphenol (3IPP), also known as m-isopropylphenol, is an alkylphenol. Its toxic impact on aquatic life has been assessed. The gasification of 3-IPP with the help of supported noble metal catalysts in supercritical water at different values of water density has been investigated. Its molar enthalpy of vaporization has been evaluated.

Application

3-Isopropylphenol (m-Isopropylphenol) may be used in the linear solvation energy relationship (LSER) studies during the characterization of stationary phases in subcritical fluid chromatography.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gasification of alkylphenols with supported noble metal catalysts in supercritical water.
Sato T, et al.
Industrial & Engineering Chemistry Research, 42(19), 4277-4282 (2003)
Characterisation of stationary phases in subcritical fluid chromatography with the solvation parameter model IV: aromatic stationary phases.
West C and Lesellier E
Journal of Chromatography A, 1115(1), 233-245 (2006)
Characterisation of stationary phases in subcritical fluid chromatography with the solvation parameter model: III. Polar stationary phases.
West C and Lesellier E
Journal of Chromatography A, 1110(1), 200-213 (2006)
Vapour pressures and enthalpies of vaporization of alkylphenols.
Nesterova TN, et al.
The Journal of Chemical Thermodynamics, 22(4), 365-377 (1990)
Stjepan Krcmar
Journal of vector ecology : journal of the Society for Vector Ecology, 32(2), 188-192 (2008-02-12)
The attraction of female tabanids to unbaited and single-baited canopy traps using 4-methylphenol, 3-isopropylphenol, and naphthalene was studied in three forest localities in eastern Croatia. Tabanids were collected in a significantly higher number in traps baited with these chemicals compared

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service