Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

479594

Sigma-Aldrich

Boc-D-phenylalaninol

98%

Synonym(s):

(R)-(+)-2-(tert-Butoxycarbonylamino)-3-phenyl-1-propanol, (R)-2-(Boc-amino)-3-phenyl-1-propanol, N-(tert-Butoxycarbonyl)-D-phenylalaninol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH[NHCO2C(CH3)3]CH2OH
CAS Number:
Molecular Weight:
251.32
Beilstein:
4686134
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

optical activity

[α]23/D +24°, c = 1 in chloroform

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

95-98 °C (lit.)

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N[C@@H](CO)Cc1ccccc1

InChI

1S/C14H21NO3/c1-14(2,3)18-13(17)15-12(10-16)9-11-7-5-4-6-8-11/h4-8,12,16H,9-10H2,1-3H3,(H,15,17)/t12-/m1/s1

InChI key

LDKDMDVMMCXTMO-GFCCVEGCSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service