Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

478989

Sigma-Aldrich

2-Isopropylphenyl isocyanate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHC6H4NCO
CAS Number:
Molecular Weight:
161.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

refractive index

n20/D 1.524 (lit.)

bp

66-67 °C/5 mmHg (lit.)

density

1.019 g/mL at 25 °C (lit.)

SMILES string

CC(C)c1ccccc1N=C=O

InChI

1S/C10H11NO/c1-8(2)9-5-3-4-6-10(9)11-7-12/h3-6,8H,1-2H3

InChI key

MWBSQVPLYJZPTG-UHFFFAOYSA-N

General description

2-Isopropylphenyl isocyanate, also known as 1-isocyanato-2-isopropylbenzene, is an aryl isocyanate.

Application

2-Isopropylphenyl isocyanate has been used in the preparation of ortho-substituted phenylcarbamate derivative of amylose.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Shen et al.
Journal of chromatography. A, 1286, 41-46 (2013-03-20)
Six ortho- and six meta-substituted phenylcarbamate derivatives of amylose were prepared and their chiral recognition abilities were evaluated as chiral stationary phases (CSPs) for high-performance liquid chromatography (HPLC). The substitution at the meta-position on the aromatic ring was more preferable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service