Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

478970

Sigma-Aldrich

4-Phenoxyphenyl isocyanate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OC6H4NCO
CAS Number:
Molecular Weight:
211.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.595 (lit.)

bp

170 °C/3 mmHg (lit.)

density

1.169 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

O=C=Nc1ccc(Oc2ccccc2)cc1

InChI

1S/C13H9NO2/c15-10-14-11-6-8-13(9-7-11)16-12-4-2-1-3-5-12/h1-9H

InChI key

PNBUGOFIKAHZRW-UHFFFAOYSA-N

General description

4-Phenoxyphenyl isocyanate, also known as 1-isocyanato-4-phenoxybenzene, is an aryl isocyanate. Its enthalpy of vaporization at boiling point has been reported.

Application

4-Phenoxyphenyl isocyanate may be used in the preparation of 1-(4-phenoxyphenyl)-3-(thiazol-2-yl)urea.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Faizul Azam et al.
Medicinal chemistry (Shariqah (United Arab Emirates)), 8(6), 1057-1068 (2012-06-30)
Synthesis of 1-(substituted aryl)-3-(thiazol-2-yl)urea derivatives was undertaken as our efforts to discover novel antiparkinsonian agents with improved pharmacological profile in haloperidol-induced catalepsy and oxidative stress in mice. Furfuryl, 2- and/or 3-methoxy substituted phenyl derivatives emerged as potent agents. With exception
Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 605-605 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service