Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

478679

Sigma-Aldrich

3-Iodophenyl isocyanate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
IC6H4NCO
CAS Number:
Molecular Weight:
245.02
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay:
97%

Assay

97%

refractive index

n20/D 1.632 (lit.)

bp

55 °C/0.2 mmHg (lit.)

density

1.86 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Ic1cccc(c1)N=C=O

InChI

1S/C7H4INO/c8-6-2-1-3-7(4-6)9-5-10/h1-4H

InChI key

LZSQANBVKPXBLO-UHFFFAOYSA-N

General description

3-Iodophenyl isocyanate, also known as 1-iodo-3-isocyanatobenzene, is a mono-iodo derivatization reagent. The derivatization of phenylhydrazine using 3-iodophenyl isocyanate during the quantification of residual phenylhydrazine in antipyrine by LC-ICP-MS (liquid chromatography-inductively coupled plasma-mass spectroscopy) has been reported.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sensitive Quantitation of Residual Phenylhydrazine in Antipyrine by LC-ICP-MS with Iodo Derivatization.
Harigaya K, et al.
Analytical Sciences, 30(8), 845-850 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service