Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

478334

Sigma-Aldrich

4-Methyl-2-nitrophenyl isocyanate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)NCO
CAS Number:
Molecular Weight:
178.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

mp

57-61 °C (lit.)

storage temp.

2-8°C

SMILES string

Cc1ccc(N=C=O)c(c1)[N+]([O-])=O

InChI

1S/C8H6N2O3/c1-6-2-3-7(9-5-11)8(4-6)10(12)13/h2-4H,1H3

InChI key

RTXDHIPQKZLDJD-UHFFFAOYSA-N

General description

4-Methyl-2-nitrophenyl isocyanate, also known as 1-isocyanato-4-methyl-2-nitrobenzene, is an organic building block containing an isocyanate group. The conformational analysis of 4-methyl-2-nitrophenyl isocyanate has been reported based on its vibrational spectral data and density functional theory (DFT) calculations.

Application

4-Methyl-2-nitrophenyl isocyanate may be used in the preparation 4-methylphenyl analog of 2-[(2-nitrophenyl)amino]furanone.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of some novel 1,3-dihydro-2H-benzimidazol-2-ylidenes.
Mack RA, et al.
The Journal of Organic Chemistry, 58(22), 6158-6162 (1993)
Vibration and DFT analysis of 2-methyl-3-nitrophenyl isocyanate and 4-methyl-2-nitrophenyl isocyanate.
Tonannavar J, et al.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 95, 120-127 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service