Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

477818

Sigma-Aldrich

2-(4-Bromophenoxy)tetrahydro-2H-pyran

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13BrO2
CAS Number:
Molecular Weight:
257.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

56-58 °C (lit.)

functional group

bromo
ether

SMILES string

Brc1ccc(OC2CCCCO2)cc1

InChI

1S/C11H13BrO2/c12-9-4-6-10(7-5-9)14-11-3-1-2-8-13-11/h4-7,11H,1-3,8H2

InChI key

MXDQGXMBJCGRCB-UHFFFAOYSA-N

General description

2-(4-Bromophenoxy)tetrahydro-2H-pyran is a halogenated heterocycle that is used as a building block in organic synthesis.

Application

2-(4-Bromophenoxy)tetrahydro-2H-pyran may be used as a starting material in the synthesis of 2-(4-lithiophenoxy)- tetrahydro-2H-pyran[1] and 4-(tetrahydropyranyloxymethyl)phenyl magnesium bromide.[2]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Fast-Moving Copper-Based Molecular Shuttle: Synthesis and Dynamic Properties.
Durola F, et al.
Chemistry (Weinheim An Der Bergstrasse, Germany), 15(16), 4124-4134 (2009)
End-group fidelity in nitroxide-mediated living free-radical polymerizations.
Rodlert M, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 38(S1), 4749-4763 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service