Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

477699

Sigma-Aldrich

Poly(styrene-alt-maleic anhydride), partial methyl ester

average Mw ~350,000

Synonym(s):

maleated copolymer, maleated polymer, maleic copolymer

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

mol wt

average Mw ~350,000

transition temp

Tg 240-250 °C

density

1.27 g/mL at 25 °C (lit.)

InChI

1S/C8H8.C5H6O4.C4H2O3/c1-2-8-6-4-3-5-7-8;1-9-5(8)3-2-4(6)7;5-3-1-2-4(6)7-3/h2-7H,1H2;2-3H,1H3,(H,6,7);1-2H/b;3-2-;

InChI key

KTCBAPXXHQDEER-PMOSZIESSA-N

Analysis Note

Contains 10-15% monomethyl ester.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service