Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

477494

Sigma-Aldrich

2,6-Bis[(4R)-(+)-isopropyl-2-oxazolin-2-yl]pyridine

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H23N3O2
CAS Number:
Molecular Weight:
301.38
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

optical activity

[α]20/D +118°, c = 0.7 in methylene chloride

mp

154-156 °C (lit.)

functional group

ether

SMILES string

CC(C)[C@@H]1COC(=N1)c2cccc(n2)C3=N[C@@H](CO3)C(C)C

InChI

1S/C17H23N3O2/c1-10(2)14-8-21-16(19-14)12-6-5-7-13(18-12)17-20-15(9-22-17)11(3)4/h5-7,10-11,14-15H,8-9H2,1-4H3/t14-,15-/m0/s1

InChI key

CSGQGLBCAHGJDR-GJZGRUSLSA-N

Application

C2 symmetric ligand for enantioselective catalysis. Easily forms bidentate coordination complexes due to the strong affinity of the oxazoline nitrogen for various metals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iryna D Alshakova et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(50), 11734-11744 (2019-07-19)
This work unveils the reactivity patterns, as well as ligand and additive effect on alkali-metal-base-catalyzed transfer hydrogenation of ketones. Crucially to this reactivity is the presence of a Lewis acid (alkali cation), as opposed to a simple base effect. With
Pfaltz, A.
Accounts of Chemical Research, 26, 339-339 (1993)
Bolm, C.
Angewandte Chemie (International Edition in English), 30, 542-542 (1991)
Gant, T.G. Meyers, A.I.
Tetrahedron, 50, 2297-2297 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service