Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

477451

Sigma-Aldrich

Triethanolamine aluminate barium salt

Synonym(s):

[μ3-[[2,2′-[[2-(Hydroxy-κO)ethyl]imino-κN]bis[ethanolato-κO:κO]](2-)]]bis[[[2,2′,2′′-(nitrilo-κN)tris[ethanolato-κO]](3-)]aluminum]barium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H37Al2BaN3O9
CAS Number:
Molecular Weight:
630.79
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

core: barium
reagent type: catalyst

SMILES string

[Ba++].OCCN(CCO[Al-]12OCCN(CCO1)CCO2)CCO[Al-]34OCCN(CCO3)CCO4

InChI

1S/C6H13NO3.2C6H12NO3.2Al.Ba/c3*8-4-1-7(2-5-9)3-6-10;;;/h8H,1-6H2;2*1-6H2;;;/q-2;2*-3;2*+3;+2

InChI key

LMRQYWULUTVMDD-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service