Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

47723

Sigma-Aldrich

N-Formylglycine

≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCONHCH2COOH
CAS Number:
Molecular Weight:
103.08
Beilstein:
1749108
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0%

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

mp

149-151 °C

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

OC(=O)CNC=O

InChI

1S/C3H5NO3/c5-2-4-1-3(6)7/h2H,1H2,(H,4,5)(H,6,7)

InChI key

UGJBHEZMOKVTIM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Kumar et al.
Journal of enzyme inhibition, 13(5), 369-376 (1998-10-30)
N-formylglycine was developed as a dead-end inhibitor of the succinic semialdehyde dehydrogenase reaction. At 4 mM, it inhibited Aspergillus niger succinic semialdehyde dehydrogenase by 40%. N-formylglycine is a reversible, complete inhibitor; the inhibition is competitive with succinic semialdehyde and uncompetitive
Einar Sagstuen et al.
The journal of physical chemistry. A, 110(28), 8653-8662 (2006-07-14)
Single crystals of the 1:1 complex of the nucleic acid base cytosine and the dipeptide N-formylglycine (C.NFG) have been irradiated at 10 and 273 K to doses of about 70 kGy and studied at temperatures between 10 and 293 K
Brian L Carlson et al.
The Journal of biological chemistry, 283(29), 20117-20125 (2008-04-09)
Type I sulfatases require an unusual co- or post-translational modification for their activity in hydrolyzing sulfate esters. In eukaryotic sulfatases, an active site cysteine residue is oxidized to the aldehyde-containing C(alpha)-formylglycine residue by the formylglycine-generating enzyme (FGE). The machinery responsible
Annette Sievers et al.
Proceedings of the National Academy of Sciences of the United States of America, 101(21), 7897-7901 (2004-05-14)
To determine the effectiveness of the ribosome as a catalyst, we compared the rate of uncatalyzed peptide bond formation, by the reaction of the ethylene glycol ester of N-formylglycine with Tris(hydroxymethyl)aminomethane, with the rate of peptidyl transfer by the ribosome.
Stefanie Jonas et al.
Journal of molecular biology, 384(1), 120-136 (2008-09-17)
The alkaline phosphatase superfamily comprises a large number of hydrolytic metalloenzymes such as phosphatases and sulfatases. We have characterised a new member of this superfamily, a phosphonate monoester hydrolase/phosphodiesterase from Rhizobium leguminosarum (R/PMH) both structurally and kinetically. The 1.42 A

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service