About This Item
Recommended Products
Assay
98%
refractive index
n20/D 1.502 (lit.)
bp
174 °C (lit.)
density
0.987 g/mL at 25 °C (lit.)
SMILES string
COCc1ccccc1
InChI
1S/C8H10O/c1-9-7-8-5-3-2-4-6-8/h2-6H,7H2,1H3
InChI key
GQKZBCPTCWJTAS-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
131.9 °F - closed cup
Flash Point(C)
55.5 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service