Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47637

Sigma-Aldrich

Fmoc-Trp-OH

≥97.0% (HPLC)

Synonym(s):

Fmoc-L-tryptophan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H22N2O4
CAS Number:
Molecular Weight:
426.46
Beilstein:
4216624
EC Number:
MDL number:
UNSPSC Code:
12352200
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Assay

≥97.0% (HPLC)

optical activity

[α]20/D −29±1°, c = 1% in DMF

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

mp

182-185 °C (lit.)

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)[C@H](Cc1c[nH]c2ccccc12)NC(=O)OCC3c4ccccc4-c5ccccc35

InChI

1S/C26H22N2O4/c29-25(30)24(13-16-14-27-23-12-6-5-7-17(16)23)28-26(31)32-15-22-20-10-3-1-8-18(20)19-9-2-4-11-21(19)22/h1-12,14,22,24,27H,13,15H2,(H,28,31)(H,29,30)/t24-/m0/s1

InChI key

MGHMWKZOLAAOTD-DEOSSOPVSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Chloé Guilbaud-Chéreau et al.
ACS applied materials & interfaces, 11(14), 13147-13157 (2019-03-14)
Molecular gels formed by the self-assembly of low-molecular-weight gelators have received increasing interest because of their potential applications in drug delivery. In particular, the ability of peptides and amino acids to spontaneously self-assemble into three-dimensional fibrous network has been exploited
Devi Wahyuningtyas et al.
Chembiochem : a European journal of chemical biology, 20(16), 2133-2140 (2019-06-06)
Huntington's disease (HD) is classified as a protein-misfolding disease correlated with the mutant Huntingtin (mHtt) protein with abnormally expanded polyglutamine (polyQ) domains. Because no effective drugs have yet been reported, attempts to develop better therapy to delay the age of
Chiao-Ting Yen et al.
European journal of medicinal chemistry, 44(5), 1933-1940 (2008-12-27)
Twenty-four new dipeptide analogs (1-24) of aurantiamide acetate were designed, synthesized, and assayed for effects on superoxide anion generation and elastase release by human neutrophils in response to fMLP/CB. Among them, seven N-(fluorenyl-9-methoxycarbonyl) (Fmoc)-dipeptides (6, 9, 12, 14, 17, 18

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service