Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

476315

Sigma-Aldrich

4,4′-Diaminobenzanilide

98%

Synonym(s):

4,4′-Diaminodiphenylamide, 4-Amino-N -(4-aminophenyl)benzamide, 4-Aminobenzoyl-4′-aminoanilide, N -(4-Aminophenyl)-4-aminobenzamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4CONHC6H4NH2
CAS Number:
Molecular Weight:
227.26
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

mp

205-207 °C (lit.)

SMILES string

Nc1ccc(NC(=O)c2ccc(N)cc2)cc1

InChI

1S/C13H13N3O/c14-10-3-1-9(2-4-10)13(17)16-12-7-5-11(15)6-8-12/h1-8H,14-15H2,(H,16,17)

InChI key

XPAQFJJCWGSXGJ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Evaluation of mutagenic activity of some chemical combinations].
E V Kleĭmenova et al.
Gigiena i sanitariia, (11-12)(11-12), 65-67 (1992-11-01)
G Della Porta et al.
Cancer letters, 14(3), 329-336 (1981-12-01)
4,4'-Diaminobenzanilide and 4,4'-diaminoazobenzene were administered in the diet to BALB/c mice at 100, 300 and 600 ppm dose levels for 60 weeks. The animals were kept under observation until 140 weeks of age when the experiment was terminated. Control and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service