Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

476102

Sigma-Aldrich

3-Pyridyl isothiocyanate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4N2S
CAS Number:
Molecular Weight:
136.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.665 (lit.)

bp

231-233 °C (lit.)

density

1.22 g/mL at 25 °C (lit.)

SMILES string

S=C=Nc1cccnc1

InChI

1S/C6H4N2S/c9-5-8-6-2-1-3-7-4-6/h1-4H

InChI key

VMSZFBSYWXMXRF-UHFFFAOYSA-N

General description

3-Pyridyl isothiocyanate (Py NCS) is an iothiocyanate derivative. Its enthalpy of vaporization at boiling point (505.15K) is 44.379kjoule/mol.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 522-522 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service