Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47548

Sigma-Aldrich

4-[2-(Fmoc-amino)ethyl]piperazin-1-ylacetic acid dihydrochloride

≥98.0% (HPLC)

Synonym(s):

4-[2-(Fmoc-amino)ethyl]-1-piperazineacetic acid dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H27N3O4 · 2HCl
CAS Number:
Molecular Weight:
482.40
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.26

Quality Level

Assay

≥98.0% (HPLC)

form

solid

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

Cl.Cl.OC(=O)CN1CCN(CCNC(=O)OCC2c3ccccc3-c4ccccc24)CC1

InChI

1S/C23H27N3O4.2ClH/c27-22(28)15-26-13-11-25(12-14-26)10-9-24-23(29)30-16-21-19-7-3-1-5-17(19)18-6-2-4-8-20(18)21;;/h1-8,21H,9-16H2,(H,24,29)(H,27,28);2*1H

InChI key

YPIAQRIOWDEWLP-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service