Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

475297

Sigma-Aldrich

3,4,6-Trimethoxy-1(3H)-isobenzofuranone

97%

Synonym(s):

3,4,6-Trimethoxyphthalide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12O5
CAS Number:
Molecular Weight:
224.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

149-152 °C (lit.)

SMILES string

COC1OC(=O)c2cc(OC)cc(OC)c12

InChI

1S/C11H12O5/c1-13-6-4-7-9(8(5-6)14-2)11(15-3)16-10(7)12/h4-5,11H,1-3H3

InChI key

JNVLFUCEKYADLU-UHFFFAOYSA-N

General description

3,4,6-Trimethoxy-1(3H)-isobenzofuranone is also referred to as 3,4,6-trimethoxyphthalide.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service