Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

475165

Sigma-Aldrich

Bis(pentamethylcyclopentadienyl)chromium(II)

96%

Synonym(s):

Decamethylchromocene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cr(C5(CH3)5)2
CAS Number:
Molecular Weight:
322.45
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

96%

form

powder or crystals

reaction suitability

core: chromium

storage temp.

−20°C

SMILES string

[Cr].C[C]1[C](C)[C](C)[C](C)[C]1C.C[C]2[C](C)[C](C)[C](C)[C]2C

InChI

1S/2C10H15.Cr/c2*1-6-7(2)9(4)10(5)8(6)3;/h2*1-5H3;

InChI key

GEYKKJXGTKHSDI-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service