Skip to Content
MilliporeSigma
All Photos(1)

Documents

474649

Sigma-Aldrich

Methyl 2-formyl-3,5-dimethoxybenzoate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOC6H2(OCH3)2CO2CH3
CAS Number:
Molecular Weight:
224.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

107-110 °C (lit.)

SMILES string

COC(=O)c1cc(OC)cc(OC)c1C=O

InChI

1S/C11H12O5/c1-14-7-4-8(11(13)16-3)9(6-12)10(5-7)15-2/h4-6H,1-3H3

InChI key

HLWARVBOEGPPGB-UHFFFAOYSA-N

Application

Methyl 2-formyl-3,5-dimethoxybenzoate may be used in the preparation of methyl (E)-3,5-dimethoxy-2-{[2-(4-methoxybenzoyl)hydrazin-1-ylidene]methyl}-benzoate.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Methyl (E)-3, 5-dimethoxy-2-{[2-(4-methoxybenzoyl) hydrazin-1-ylidene] methyl} benzoate.
Naz H, et al.
Acta Crystallographica Section E, Structure Reports Online, 68(9), o2671-o2671 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service