Skip to Content
MilliporeSigma
All Photos(1)

Documents

473871

Sigma-Aldrich

Ethyl difluoroiodoacetate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5F2IO2
CAS Number:
Molecular Weight:
249.98
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

Assay

95%

contains

copper as stabilizer

SMILES string

CCOC(=O)C(F)(F)I

InChI

1S/C4H5F2IO2/c1-2-9-3(8)4(5,6)7/h2H2,1H3

InChI key

HGWXXIZVRRTDKT-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service