Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

473766

Sigma-Aldrich

(R)-(+)-3-Benzyloxy-2-(Boc-amino)-1-propanol

98%

Synonym(s):

(R)-(+)-3-Benzyloxy-2-(tert-butoxycarbonylamino)-1-propanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OCH2CH[NHCO2C(CH3)3]CH2OH
CAS Number:
Molecular Weight:
281.35
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

optical activity

[α]20/D +14°, c = 1 in chloroform

mp

66-69 °C (lit.)

functional group

amine
ether
hydroxyl
phenyl

SMILES string

CC(C)(C)OC(=O)N[C@H](CO)COCc1ccccc1

InChI

1S/C15H23NO4/c1-15(2,3)20-14(18)16-13(9-17)11-19-10-12-7-5-4-6-8-12/h4-8,13,17H,9-11H2,1-3H3,(H,16,18)/t13-/m1/s1

InChI key

MSIDLARYVJJEQY-CYBMUJFWSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service