Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47313

Sigma-Aldrich

Fmoc-D-Cha-OH

≥98.0% (TLC)

Synonym(s):

Fmoc-3-cyclohexyl-D-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H27NO4
CAS Number:
Molecular Weight:
393.48
Beilstein:
8234680
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98.0% (TLC)

storage temp.

2-8°C

SMILES string

OC(=O)[C@@H](CC1CCCCC1)NC(=O)OCC2c3ccccc3-c4ccccc24

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoichiro Yamada et al.
Bioorganic & medicinal chemistry, 19(1), 168-171 (2010-12-21)
High mobility group box1 (HMGB1) is a non-histone chromatin chromosomal protein playing an important role in chromatin architecture and transcriptional regulation. Recently, HMGB1 has been shown to be secreted into extracellular milieu in necrosis and apoptosis, and involved in inflammatory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service