Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

472824

Sigma-Aldrich

4-O-Acetyl-3,6-di-O-(tert-butyldimethylsilyl)-D-glucal

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H40O5Si2
CAS Number:
Molecular Weight:
416.70
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

97%

optical activity

[α]20/D −19°, c = 2.1 in chloroform

refractive index

n20/D 1.456 (lit.)

bp

324 °C (lit.)

density

0.968 g/mL at 25 °C (lit.)

SMILES string

CC(=O)O[C@@H]1[C@@H](CO[Si](C)(C)C(C)(C)C)OC=C[C@H]1O[Si](C)(C)C(C)(C)C

InChI

1S/C20H42O7Si2/c1-14(22)26-17(16(24)13-25-28(8,9)19(2,3)4)18(15(23)12-21)27-29(10,11)20(5,6)7/h12,15-18,23-24H,13H2,1-11H3/t15-,16+,17+,18+/m0/s1

InChI key

PUTBIZQBKYLDAU-BSDSXHPESA-N

Looking for similar products? Visit Product Comparison Guide

Application

Important building block for both solution- and solid-phase synthesis of oligosaccharides.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seeberger, P.H., et al.
Aldrichimica Acta, 30, 75-75 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service