Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

472719

Sigma-Aldrich

Ethyl (R)-nipecotate, L-tartrate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H21NO8
CAS Number:
Molecular Weight:
307.30
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]20/D +10°, c = 5 in H2O

mp

157-159 °C (lit.)

SMILES string

O[C@H]([C@@H](O)C(O)=O)C(O)=O.CCOC(=O)[C@@H]1CCCNC1

InChI

1S/C8H15NO2.C4H6O6/c1-2-11-8(10)7-4-3-5-9-6-7;5-1(3(7)8)2(6)4(9)10/h7,9H,2-6H2,1H3;1-2,5-6H,(H,7,8)(H,9,10)/t7-;1-,2-/m11/s1

InChI key

HHPGQKZOPPDLNH-NWAAXCJESA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service