Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

471755

Sigma-Aldrich

8-Hydroxyquinoline zinc

99%

Synonym(s):

Bis-(8-hydroxyquinolinato)zinc, Zinc 8-hydroxyquinolinate, Zinc oxinate, Znq

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12N2O2Zn
CAS Number:
Molecular Weight:
353.69
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

mp

>350 °C (lit.)

λmax

255 nm

fluorescence

λex 325 nm; λem 484 nm in chloroform

OLED Device Performance

ITO/TPD/Znq/Mg:In

  • Color: yellow
  • Max. Luminance: 16200 Cd/m2

SMILES string

C[Zn](C)(Oc1cccc2cccnc12)Oc3cccc4cccnc34

InChI

1S/2C9H7NO.Zn/c2*11-8-5-1-3-7-4-2-6-10-9(7)8;/h2*1-6,11H;/q;;+2/p-2

InChI key

HTPBWAPZAJWXKY-UHFFFAOYSA-L

General description

Bis(8-hydroxyquinoline) zinc(II) complex (Znq2), an electroluminescent material.

Application

Bis(8-quinolinolato-N1,O8)-zinc(II) complex (Znq2) can be employed as an electron transfer layer in organic light-emitting diodes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

In situ formation of bis(8-hydroxyquinoline) zinc(II) complex in the interlayer spaces of smectites by solid?solid reactions
Khaorapapong N and Ogawa M
Journal of Physics and Chemistry of Solids null
In situ formation of bis(8-hydroxyquinoline) zinc(II) complex in the interlayer spaces of smectites by solid?solid reactions
Khaorapapong N and Ogawa M
Journal of Physics and Chemistry of Solids, 941-948 null
Quinolinate zinc complexes as electron transporting layers in organic light-emitting diodes
Donze N, et al.
Chemical Physics Letters null
Organic Electroluminescent Devices with 8-Hydroxyquinoline Derivative-Metal Complexes as an Emitter
Hamada, Y.; et al.
Japanese Journal of Applied Physics, 32, L514-L515 (1993)
Quinolinate zinc complexes as electron transporting layers in organic light-emitting diodes
Donze N, et al.
Chemical Physics Letters null

Articles

Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service