Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

471186

Sigma-Aldrich

2-Pentanone

spectrophotometric grade, ≥99%

Synonym(s):

Methyl propyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2CH2CH3
CAS Number:
Molecular Weight:
86.13
Beilstein:
506058
EC Number:
MDL number:
UNSPSC Code:
12190000

grade

spectrophotometric grade

vapor pressure

27 mmHg ( 20 °C)

Assay

≥99%

autoignition temp.

941 °F

expl. lim.

8.2 %

impurities

<0.050% water

evapn. residue

<0.0005%

refractive index

n20/D 1.39 (lit.)

bp

101-105 °C (lit.)

mp

−78 °C (lit.)

density

0.809 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 330 nm Amax: 1.00
λ: 340 nm Amax: 0.10
λ: 350 nm Amax: 0.01
λ: 370-400 nm Amax: 0.005

SMILES string

CCCC(C)=O

InChI

1S/C5H10O/c1-3-4-5(2)6/h3-4H2,1-2H3

InChI key

XNLICIUVMPYHGG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

46.4 °F - closed cup

Flash Point(C)

8 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shane Avison et al.
Journal of agricultural and food chemistry, 63(28), 6313-6318 (2015-06-23)
A new methodology is presented to measure water-air partition coefficients (Henry's constants) of volatiles, using APCI-MS. Significant advantages over other Henry's constant determination methods include the short measurement and sample preparation time and the possibility for simultaneous measurement of multiple
Giulia Caron et al.
Journal of medicinal chemistry, 48(9), 3269-3279 (2005-04-29)
Growing interest in the use of both the logarithm of the partition coefficient of the neutral species in the alkane/water system (log P(N)(alk)) and the difference between log P(N)(oct) (the logarithm of the partition coefficient of the neutral species in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service