Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

471151

Sigma-Aldrich

5,12-Bis(phenylethynyl)naphthacene

technical grade, 85%

Synonym(s):

BPEN

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H20
CAS Number:
Molecular Weight:
428.52
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

grade

technical grade

Assay

85%

mp

248 °C (dec.) (lit.)

λmax

550 nm

SMILES string

c1ccc(cc1)C#Cc2c3ccccc3c(C#Cc4ccccc4)c5cc6ccccc6cc25

InChI

1S/C34H20/c1-3-11-25(12-4-1)19-21-31-29-17-9-10-18-30(29)32(22-20-26-13-5-2-6-14-26)34-24-28-16-8-7-15-27(28)23-33(31)34/h1-18,23-24H

InChI key

OUHYGBCAEPBUNA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

An n-channel organic semiconductor.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Newman, C.R., et al.
Chemistry of Materials, 16, 4436-4436 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service